Eisco Light Wall Glass Test Tubes with Beaded Rim
Supplier: Wards
CH0718APK48 CH0718BPK48 CH0718CPK48 CH0718D01PK48 CH0718EPK48 CH0718FPK24 CH0718F01PK24 CH0718GPK24 CH0718HPK24 CH0718AAPK48
470323-142PK 29.5 CAD
470323-142 470323-144 470323-146 470323-148 470323-150 470323-152 470323-154 470323-156 470323-158 470323-166
Eisco Light Wall Glass Test Tubes with Beaded Rim
Tubes Test Tubes
Light wall test tube with beaded rim for general use.
  • Made from high quality Borosilicate 3.3 glass
  • Feature fire polished, beaded rim, and uniform wall thickness
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient- can withstand direct flame, high temperatures and thermal shocks
Eisco Labs Borosilicate glass has the following heat tolerances: strain point –515 °C; annealing point 565 °C; softening point 820 °C. They manufacture their own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.