Eisco Porcelain Evaporating Basins, Round Form
Supplier: Wards
CH0071A CH0071B CH0071C CH0071D CH0071E CH0071F CH0071G CH0071H
470328-268EA 3.65 CAD
470328-268 470328-270 470328-272 470328-274 470328-276 470328-278 470328-280 470328-282
Eisco Porcelain Evaporating Basins, Round Form
Trays/Dishes/Pots Evaporating Basins/Dishes
Designed for a variety of science laboratory uses.
  • High quality, fully glazed except outer base
  • White porcelain
  • Round form with spout
  • Heat and cool slowly when using under high temperatures
Eisco Labs manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.