Nichipet Air Fully Autoclavable Micro Pipettes, Nichiryo America
Supplier: Nichiryo America
00-NAR-2 00-NAR-10 00-NAR-20 00-NAR-100 00-NAR-200 00-NAR-1000 00-NAR-5000 00-NAR-10000 00-NAR-100M12 00-NAR-100M8 00-NAR-10M12 00-NAR-10M8 00-NAR-200M12 00-NAR-200M8 00-NAR-300M12 00-NAR-300M8
76457-084EA 541.94 CAD
76457-084 76457-086 76457-088 76457-090 76457-092 76457-094 76457-096 76457-098 76637-132 76637-140 76637-142 76637-138 76637-144 76637-134 76637-136 76637-146
Nichipet Air Fully Autoclavable Micro Pipettes, Nichiryo America
Pipettes Micropipettes
Lightest weight, lighter stroke resistance, 8 different colors and easy calibration while maintaining higher accuracy and precision, and fully autoclaveble feature.
  • Only 73.5 g (1000 µl)
  • Easy handling and volume setting
  • Easy self-calibration
  • Longer second push (2 µl, 10 µl and 20 µl)
  • Ergonomic shape
  • Easy recognition (each volume sizes in different color)
  • Filtered nozzles are used for large volume models (5000 µl and 10000 µl)
Available in eight different sizes covering a volume range of 0.1 µl to 10 ml, pipettors made of UV resistance material and lightest weight and lighter push resistance to reduce fatigue. A smooth, continuous, and adjustable micrometer replaces standard click-stop versions, allowing desired volumes to be dialed in for precise measurement. The 5000 µl, and 10 ml models are equipped with filtered nozzles.
Certifications: Test Report comply with ISO8655-6 standard is eclosed each pipette.
Caution: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.