Bovine Deoxyribonuclease I, RNase- and Protease-Free (from Pancreas)

Supplier: Rockland Immunochemical

MB-101-0100 MB-101-0500
CAMB-101-0100EA 188 CAD
CAMB-101-0100 CAMB-101-0500
Bovine Deoxyribonuclease I, RNase- and Protease-Free (from Pancreas)
Enzymes

All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.


Application includes splitting phosphodiester linkages to yield polynucleotides with a Free 3'-OH and 5'-Phosphate.

Order Now

SPECIFICATIONS

Enzyme name Deoxyribonuclease I, RNase- and Protease-Free
Environmentally Preferable
Source Pancreas
Species Bovine
Protease-free Y

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR