VWR® Bovine Ribonuclease A (from Pancreas)

Supplier: VWR

E866-1ML E866-5ML
97063-062EA 102.13 CAD
97063-062 97064-064
VWR® Bovine Ribonuclease A (from Pancreas)
Enzymes

Ribonuclease A is an endoribonuclease that efficiently hydrolyzes RNA contaminants in DNA preparations from tissue or bacterial cell cultures. RNase A is used for a variety of molecular biology applications, most commonly for the hydrolysis of RNA that contaminates DNA preparations. This enzyme is used during plasmid purification without nicking or degrading plasmid.


  • Useful during DNA purification
  • High purity
  • Available in a ready-to-use solution


Ribonuclease A is an endoribonuclease that efficiently hydrolyzes RNA contaminants in DNA preparations from tissue or bacterial cell cultures. RNase A is used for a variety of molecular biology applications, most commonly for the hydrolysis of RNA that contaminates DNA preparations. This enzyme is used during plasmid purification without nicking or degrading plasmid.

Order Now

SPECIFICATIONS

Enzyme name Ribonuclease A
Environmentally Preferable
Source Pancreas
Species Bovine
Form Lyophilized powder
Enzyme activity 10 mg/ml
pH pH 8.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR