L-Threonine 98.5-101.5% (dried basis) USP, Multi-Compendial, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

MACR4947-06 MACR4947-07
CAMK4947-06EA 5961.94 CAD
CAMK4947-06 CAMK4947-07
L-Threonine 98.5-101.5% (dried basis) USP, Multi-Compendial, Macron Fine Chemicals™
L-Threonine
Formula: C₄H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C₄H₈NO₃) (dried basis) 98.5 - 101.5 %
USP - Identification Passes Test
USP - Specific Rotation [Å]²⁵ ^D -29.1 - -26.7 Degree
USP - pH 5.0 - 6.5
USP - Loss on Drying at 105°C ≤ 0.2 %
USP - Residue on Ignition ≤ 0.4 %
USP - Chloride (Cl) ≤ 0.05 %
USP - Sulfate (SO₄) ≤ 0.03 %
USP - Iron (Fe) ≤ 0.003 %
USP - Related Compounds - Individual Impurities ≤ 0.5 %
USP - Related Compounds - Total Impurities ≤ 2.0 %
EP/BP - Assay (C₄H₈NO₃) (dried basis) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH 5.0 - 6.5
EP/BP - Specific Rotation [Å]²º^D -29.0 - -27.6 Degree
EP/BP-Ninhydrin-Positive Substances-Each ≤ 0.2 %
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤ 0.5 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Ammonium (NH₄) ≤ 0.02 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 0.5 %
EP/BP - Ash (sulfated) ≤ 0.1 %
JP - Assay (C₄H₈NO₃) (dried basis) 98.5 - 101.0 %
JP - Identification Passes Test
JP - Optical Rotation -29.0 - -26.0 Degree
JP - pH 5.2 - 6.2
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.021 %
JP - Sulfate (SO₄) ≤ 0.028 %
JP - Ammonium (NH₄) ≤ 0.02 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 2 ppm
JP - Related Substances Passes Test
JP - Loss on Drying at 105°C ≤ 0.20 %
JP - Residue on Ignition ≤ 0.10 %
Endotoxin Concentration, IU/mg, For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR