Tungsten Carbide Cutters, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
9506 9520 9500
76458-566EA 515.84 CAD
76458-566 76458-568 76458-570 76458-572 76458-574 76458-576 76458-578 76458-580 76458-582 76458-584 76458-586 76458-588 76458-590 76458-592 76458-594 76458-596 76458-598 76458-600 76458-602 76458-604 76458-606 76458-608 76458-610 76458-612 76458-614 76458-616 76458-618 76458-620 76458-622 76458-624 76458-626 76458-628 76458-630 76458-632 76458-634 76458-636
Tungsten Carbide Cutters, Electron Microscopy Sciences
Cutters
These cutters and pliers are designed for use in applications where strength, wear resistance, and hardness are a primary concern.
  • Magnetizable
  • Harden by heat treatment
  • ESD safe
Tungsten carbide and high precision cutters are made from carbon steel, with a maximum hardness of 68 HRC. They offer a moderate resistance to corrosion, and are ideal for cutting and bending single or multiple filars, and for lateral or internal cuts.
ESD epoxy-coated cutting tweezers have a poor resisance to corrosion. They are often used as tweezers in medical deviceMore Product Information