Nitrile Examination Gloves, Textured, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
71116-91 71116-92 71116-94 71116-90
76458-556BX 38.25 CAD
76458-556 76458-558 76458-560 76458-562 76458-564
Nitrile Examination Gloves, Textured, Electron Microscopy Sciences
Gloves
Gloves are made from 100% synthetic nitrile that contains no latex. They offer superior textile strength and are puncture-resistant when compared to other latex or vinyl gloves of similar thickness.
  • Textured finish provides a positive grip on wet and/or dry surfaces
  • Excellent textile strength
  • Puncture-resistant
  • Latex-free
  • Powder-free
These powder-free gloves will provide the user with superior comfort, excellent flexibility, and dexterity. The nitrile compound provides a broad range of hand protection against accidental chemical splashes. There are no natural rubber protein allergens as is present in natural latex rubber gloves.