Glassware washer accessories, FlaskScrubber® and SteamScrubber®
Supplier: Labconco
4573000 4573100 4573400 4573500 4573600 4573700 4573800 4573900 4574200 4574600 4575700 4574700 4574800 4574900 4575200 4575300 4575400 4575500 4574400 4574500 4587520 4595120 4679700 4653500 4653300 4678000 4643000 4669540 4679300 4679301 4668501 4668601 4668901 4668500 4668600 4668700 4668800 4668900 4669000 4575800 4678400 4678401 4596110 4596111 4681300 4681301
76449-150EA 361.82 CAD
76449-150 76449-152 76449-154 76449-156 76449-158 76449-160 76449-162 76449-164 76449-166 76449-168 76449-170 76449-172 76449-174 76449-176 76449-178 76449-180 76449-182 76449-184 76449-186 76449-188 76449-190 76449-192 76449-194 76449-196 76449-198 76449-200 76449-202 76449-204 76449-206 76449-208 76449-210 76449-212 76449-214 76449-216 76449-218 76449-220 76449-222 76449-224 76449-226 76449-228 76449-230 76449-232 76449-234 76449-236 76449-238 76449-240 76449-242
Glassware washer accessories, FlaskScrubber® and SteamScrubber®
Glassware Washers
There is a wide selection of accessories to customize your glassware washer to your specific needs and applications. These racks and accessories are for use with Labconco FlaskScrubber®, SteamScrubber®, FlaskScrubber® Vantage glassware washers. Racks and inserts are stainless steel unless otherwise stated.
  • Versatile inserts and specialized wash accessories
  • Additional, interchangeable racks for upper and lower positions
  • Optional accessories to make the most of your Labconco glassware washer including printers and automatic detergent dispensers
Tray insert for pans, instrument trays, multi-purpose trays and other shallow containers. Capacity: 5 trays. Utensil basket with cover for miscellaneous labware such as stoppers, spatulas and other small lightweight items. Mesh basket is a full rack insert, open mesh basket that can be used for a variety of utensils and loose labware. The bulk tube insert's quick-loading design holds up to 600 each 13 mm tubes or 244 each 20 mm tubes. Divider securely holds tubes during wash cycle. Culture tube inserts are designed to hold 10 to 12 mm, 15 to 18 mm or 20 to 24 mm Ext.Ø tubes. Retainer top included with 10 to 12 mm insert only but may be purchased separately. 48-Pin insert securely holds smaller volume wide-mouth labware. Silicone tips prevent scratching. 10-Pin and 32-Pin inserts also available. Graduated cylinder insert holds up to 3 graduated cylinders. Includes 3 full-coverage spray nozzles for maximum cleanability. Up to 2 inserts per rack.
Full-coverage spray nozzle distributes water to maximize cleanability of wide-mouth labware. Includes 1 spray nozzle and 1 spindle. Test tube holders accommodate test tubes with 10 to 13 mm, 14 to 20 mm or 20 to 25 mm Ext.Ø. Two may be stacked for increased capacity. Stacked holders can't accommodate an upper rack in the washer. Requires Labconco 4575300 conversion kit and lower spindle rack (not included). Utensil holder for spatulas, spoons, scoops and other miscellaneous utensils. Utensils fit into 6 spacious compartments. Test tube spindle conversion kit includes 6 spindle rows with 8 spindles per row and a supportive base. Requires test tube holder insert 4574700, 4574800 or 4574900. Pipette inserts securely holds up to 6 or 24 pipettes vertically to clean and dry pipettes. Vial inserts securely hold up to 48 (12×32 mm or 15×45 mm) vials during washing. Includes retainer lid with handle, support holder and 6 spindle arms. May use up to 2 per spindle rack.
Liquid detergent dispenser kit, acid rinse dispenser kit and detergent and acid rinse dispenser kit connect to any SteamScrubber® or FlaskScrubber® washer to automate liquid detergent or acid rinse dispensing, or both. Includes peristaltic pump, float switch, tubing, and either a one-gallon plastic container (detergent kit), a one-liter plastic container (acid rinse kit), or both. Detergent and acid rinse not included.
Base stands elevate freestanding SteamScrubber® and FlaskScrubber® or FlaskScrubber® Vantage washers by 17" for loading convenience. Includes one storage compartment with hinged door. Stationary stand, fully assembled with leveling feet.
Ethernet kit provides connectivity for remote updates and access to data. For use with Labconco SteamScrubber and FlaskScrubber washers with 9-digit catalog number.
Detergent storage cabinet provides convenient compartment to store automatic detergent dispense pumps and bottles for Labconco SteamScrubber® and FlaskScrubber® washers.
Protective opaque silicone tips cover the ends of spindle tips to prevent scratching of glass and metal labware. 40 silicone tips per bag.
Upper rack retrofit kit allows for use of current high-flexibility upper racks in Labconco FlaskScrubber® or SteamScrubber® washers with 7-digit catalog numbers beginning with 442 and 440. Includes retrofit slides and hardware. Installation required.
Heater cover provides barrier layer between labware and heating coils in Labconco glassware washers. Unique design provides additional filtration layer to protect from broken glass.
Small spindle racks have a high-capacity design with 60 spindles, 60 spindle plugs, 12 spindle arm plugs, spindle tool and wash arm for cleaning of small narrow-neck labware in Labconco glassware washers. Catalog number for upper rack for FlaskScrubber® and FlaskScrubber® Vantage includes upper slide rails for installation. Lower small spindle rack may be used in place of any lower rack in Labconco glassware washers.
Spindle racks are designed for narrow-neck labware in Labconco glassware washers. Includes 36 spindles, 36 adjust height clips, 18 glassware holders, 10 small spindles & clips, 36 spindle plugs, 12 spindle arm plugs. Catalog number for upper rack for FlaskScrubber® and FlaskScrubber® Vantage includes upper slide rails for installation. Lower spindle rack is included with every FlaskScrubber® and FlaskScrubber® Vantage and may be used in place of lower standard rack in SteamScrubber® washers.
Standard racks are open to accommodate accessory inserts to suit your labware needs (sold separately). Includes wash arm. May be used in any Labconco glassware washer. Catalog number for upper rack for FlaskScrubber® and FlaskScrubber® Vantage includes upper slide rails for installation. Lower standard rack is included with every SteamScrubber® and may be used in place of lower spindle rack in any FlaskScrubber® or FlaskScrubber® Vantage washer.
Interchangable 5-spindle spray arm has 5 direct-injection spindles for cleaning of small narrow-neck labware.
Drain water Ccooling kit provides plumbing components, solenoid valve and electrical connections necessary to reduce drain water temperature to <60 °C (<140 °F). Requires cold water connection. For use in Labconco glassware washers. 115 or 230 V options available. Freestanding-to-Mobile conversion kit adds mobility to freestanding washer models. Allows washers to be plugged into a 20 A electrical wall outlet and connected to a standard laboratory sink faucet. For use with Labconco FlaskScrubber® and SteamScrubber® washers. Not compatible with FlaskScrubber Vantage models. 115 or 230 V options available.
Compact thermal printer produces summary report of washer cycle. Includes USB data cable and rechargeable NiMH AA batteries. Separate models for use with Labconco FlaskScrubber® and SteamScrubber® washers or Labconco FlaskScrubber® Vantage washers.