Intelsius® BioTherm (2...8 °C) Sample Transport Shippers, DGP Intelsius
Supplier: DGP INTELSIUS LLC
BT056 BT057 BT060 BT061 BT068 BT070 BT071 BT072 BT073
76441-424EA 125.29 CAD
76441-424 76441-426 76441-428 76441-430 76441-432 76441-436 76441-438 76441-440 76441-442 76441-444
Intelsius® BioTherm (2...8 °C) Sample Transport Shippers, DGP Intelsius
Mailers and Shippers
Intelsius® BioTherm shippers are suitable for shipping temperature critical samples requiring a stable fridge temperature of between 2 and 8 °C.
  • Passive cooling
  • Compliant labels
  • Reusable cold pack technology
  • Regulatory compliant packaging
The BioTherm range of high performance 2 to 8 °C solutions are fully compliant to IATA, ADR, and DOT (49 CFR) transport regulations. These systems are qualified to ensure sample integrity for up to 96 hours against 2 to 8 °C temperature exposures. Each system is provided with handles and the correct compliance labeling to enable careful handling throughout the regulatory compliant cold chain. Cold packs are included.