Intelsius® PharmaTherm Pallet Shippers, DGP Intelsius
Supplier: DGP Intelsius
PHT230 PHTP440W PHTP90S PHTP180S PHTP90W PHTP440S PHT375 PHTP180W PHT800
76441-522EA 1215.64 CAD
76441-522 76441-532 76441-520 76441-516 76441-526 76441-524 76441-518 76441-528 76441-530
Intelsius® PharmaTherm Pallet Shippers, DGP Intelsius
Mailers and Shippers
The Intelsius® Pharmatherm pallet shipping systems are designed to offer optimal thermal protection for pharmaceutical products.
  • Large internal volume
  • Optimal thermal protection
  • Pallet sized shipper
These shipping systems are pre-qualified to maintain payload integrity during shipments for up to 96 hours. The range is tested against Intelsius® demanding high performance dry ice temperature and high performance 2 to 8 °C profiles.