Intelsius® PathoShield Complete Sample Shipping Solution Packages, DGP Intelsius
Supplier: DGP INTELSIUS LLC
PA002A PA003A PA006A PA007A
76441-506BX 249.25 CAD
76441-506 76441-508 76441-510 76441-512 76441-514
Intelsius® PathoShield Complete Sample Shipping Solution Packages, DGP Intelsius
Mailers and Shippers
The PathoShield range are complete shipping solutions designed to carry a wide range of primaries, including blood vials, swabs, and sample containers.
  • Complete sample transport solution
  • Includes 95kPa PathoPouch
  • Pre-printed with required compliant markings
Each PathoShield complete system is pre-printed with the required markings and includes the relevant components to ship samples in compliance with transport regulations. The PathoShield solutions are ideal when transporting samples for COVID-19 testing and analysis.
Certifications: CompleteMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable