Inksmith Mako Laser Cutters
Supplier: INKSMITH LIMITED SE
LSR-MAKO-40 LSR-MAKO-80
470330-324EA 10299.99 CAD
470330-324 470330-326
Inksmith Mako Laser Cutters
Cutters
Perfect classroom laser cutter.
  • Up to 0.5" cutting depth
  • Interlocking UV cover
  • Optional external Fume Filtration unit allows for the safe and clean operation of the laser cutter indoors
  • InkSmith Mako CO₂ Laser Cutter has been tested and ESA certified for use in classrooms
  • Machines are equipped with advanced safety features, making laser cutting fun and enjoyable for everyone
  • WiFi compatible
Achieve instant results on a variety of materials with the InkSmith Mako40 CO₂ Laser Cutter. The Mako Laser Cutter is a series of industry-standard laser cutting machines designed for education. The Mako is equipped and certified for safe operation in any makerspace or classroom. With large workspaces and units equipped with fume extraction and filtering, these cutters are perfect for makerspaces, classrooms, and STEM projects.