VWR® Precision Grip Gloves with Dots
Supplier: VWR International
GCVWRWRKA1DB06 GCVWRWRKA1DB07 GCVWRWRKA1DB08 GCVWRWRKA1DB09 GCVWRWRKA1DB10 GCVWRWRKA1DB11
76408-568CS 1504.38 CAD
76408-568 76408-570 76408-572 76408-574 76408-576 76408-578
VWR® Precision Grip Gloves with Dots
Gloves
The VWR® precision grip glove with grip dots and polyurethane coating provide basic cut protection and enhanced grip. Ideal for handling, picking, packaging, automotive, or situations that require dexterity in a dry and/or dirty environment.
  • Grip dots
  • Nylon composition
  • Nitrile (NBR) coated
  • Gray knit glove with black coating
Certifications: Meets requirements for ANSI A1 Cut Protection (Level A). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.