VWR® Cut Protection Gloves, NBR Coating
Supplier: VWR International
76408-520 76408-522 76408-524 76408-526 76408-528 76408-530
76408-520CS 1531.11 CAD
76408-520 76408-522 76408-524 76408-526 76408-528 76408-530
VWR® Cut Protection Gloves, NBR Coating
Gloves
The VWR® cut protection glove with nitrile coating provides general cut protection for the widest range of applications.
  • Nylon/spandex/polyester blend composition
  • Nitrile (NBR) coated
  • Gray knit glove with black coating
Ideal for use in environments handling items or tools with a risk of cuts, including material handling, assembly, maintenance, and janitorial applications.
Certifications: Meets requirements for ANSI A2 Cut Protection (Level B). EN388:2016. The CE marking on these products means that they meet the requirements of European directive 89/686/CEE or EU Regulation 2016/425 on Personal Protective Equipment concerning protection, comfort and strength. Certified by OKEO-TEX meeting Standard 100 certifying that no harmful substances are used in the production of these products.