Intelsius® Pharmachill Reusable Cold Packs, DGP Intelsius
Supplier: DGP INTELSIUS LLC
BS668 BS669 BS670 BS671
76441-400BX 231.61 CAD
76441-400 76441-402 76441-404 76441-406
Intelsius® Pharmachill Reusable Cold Packs, DGP Intelsius
Cooling Packs
Pharmachill cold packs are reusable thermal media solutions using a water-based PCM ideal for maintaining payload temeprature at between 2 to 8 ˚C and 15 to 25 ˚C.
  • Reusable
  • Durable hard plastic
  • Phase change material
Intelsius® produces a range of thermal media tailored to varied performance requirements. The Pharmachill range includes rigid, liquid-filled coolant bottles designed for use with temperature controlled packaging solutions.
Pharmachill Cold Packs feature a non-toxic, water based PCM. They have been specifically designed to be reusable,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable