Avantor Single Use 3D Tank Liners with Bottom Drain, Avantor Fluid Handling
Supplier: Avantor Fluid Handling
1003013 1003014 1003015 1003016 1003017
76452-770CS 13174.12 CAD
76452-770 76452-772 76452-774 76452-776 76452-778
Avantor Single Use 3D Tank Liners with Bottom Drain, Avantor Fluid Handling
Bags Media Bags
These tank liners are single-use bags with an open top, designed to fit in cylindrical vessels.
  • No cleaning after media preparation
  • Medical grade, multi-layer film
  • For bioprocess applications
  • Delivered gamma irradiated and ready-to-use
These bags which have a bottom drain are used as a liner into a vessel to avoid cleaning of the vessel after media and buffer preparation. After installing the Tank Liner into a vessel and filling it with liquid, powder can be added to the vessel and the mixing process can start by using an overhead agitator. WhenMore Product Information