ComfortFlex® 5 mil Powder-Free Nitrile/PVC Blend Disposable Gloves, Magid
Supplier: Magid Glove
T5360S T5360M T5360L T5360XL
76451-958CS 219.96 CAD
76451-958 76451-960 76451-962 76451-964
ComfortFlex® 5 mil Powder-Free Nitrile/PVC Blend Disposable Gloves, Magid
Gloves
Unique PVC/nitrile material blend is designed for the majority of single-use applications.
  • Lightweight nitrile/PVC blend provides a sensitive bare-hand feel
  • Rolled cuff to prevent tearing when donning the glove
  • Economical protection for applications that involve dirt, certain chemicals, and fluids
  • Excellent choice as a liner under another glove
Certifications: Complies with FDA food handling requirements (21 CFR 177.2600).
Packaging: Case of 1000 gloves.
Caution: The service life of this product will vary depending on the application and type of care administered. This product should be replaced immediately once a critical or major defect is noted.