Rotary Evaporators with Slide Jack Mechanism, Yamato

Supplier: Yamato Scientific
RE-202-AWA RE-202-BWA RE-202-AOA RE-202-BOA
76443-154KT 6657.8 CAD
76443-154 76443-156 76443-158 76443-160
Rotary Evaporators with Slide Jack Mechanism, Yamato
Evaporators Rotary Evaporators
These standard rotary evaporators feature convenient height adjustment and intuitive operation which increases user-comfort and productivity.

  • Option to use either a water bath or oil bath with a 5 L capacity
  • Experience high-impact performance
  • Market leader in rotation speed range at 5 to 315 rpm
  • Set inversion function where glassware and bath can be set on either side, left or right, depending on user’s dominant hand and installation location
  • Standard equipped with condenser insulation kit to suppress condensation on the condenser’s surface
  • Convenient
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Dimensions 744×365×534(Overall dimensions) 554×365×745((Overall dimensions)
Temperature range 5...35 °C 5...35 °C
Control range 5 – 315 rpm 5 – 315 rpm
Condenser Glass Glass

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR