VWR® Plus Series Solid Door Undercounter Built-in ADA Compliant Refrigerators
Supplier: VWR International
76422-930 76422-932
76422-930EA 2758.36 CAD
76422-930 76422-932
VWR® Plus Series Solid Door Undercounter Built-in ADA Compliant Refrigerators
Refrigerators
These undercounter refrigerators provide lab quality cold storage with ADA compliance.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft circulation
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76422-930, 76422-932
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Cat. No. 76422-930 76422-932
Defrost Cycle Cycle
Shelves/drawers 2 adjustable, 1 fixed 2 adjustable, 1 fixed
Door type One solid swing door, right hinged One solid swing door, left hinged
Electrical 115 V, 60 Hz, 1.74 A, 1/8 HP 115 V, 60 Hz, 1.74 A, 1/8 HP
Shipping weight 68 kg (150 lbs.) 68 kg (150 lbs.)
Environmental certification HFC-free R600a refrigerant HFC-free R600a refrigerant