VWR® Sterile Cleanroom Nitrile Glove, Class 100
Supplier: VWR International
76423-444 76423-446 76423-448 76423-450 76423-452 76423-454
76423-444CS 402.87 CAD
76423-444 76423-446 76423-448 76423-450 76423-452 76423-454
VWR® Sterile Cleanroom Nitrile Glove, Class 100
Gloves
Introducing the new VWR® Class 100 sterile nitrile gloves as a sterile bulk packed option to the popular line of VWR® cleanroom gloves. Enhanced comfort and durability make these an ideal choice to add to any glove program.
  • Sterile SAL 10⁻⁶, gamma irradiation
  • Nitrile
  • Ambidextrous
  • Textured fingers
  • Beaded cuff
  • Natural rubber, latex-free
Suitable for use in ISO Class 5 (FED-STD-209E Class 100) or higher cleanrooms.
Packaging: Gloves are packaged 200 per bag, 5 bags per case. Packaging is made of 40% recycled material.