High Performance FR Camo Jackets, Lakeland Industries
Supplier: Lakeland
JKTAT30-2X JKTAT30-2XT JKTAT30-3X JKTAT30-3XT JKTAT30-4X JKTAT30-4XT JKTAT30-LG JKTAT30-LGT JKTAT30-MD JKTAT30-MDT JKTAT30-SM JKTAT30-SMT JKTAT30-XL JKTAT30-XLT
76438-856EA 771.04 CAD
76438-856 76438-858 76438-860 76438-862 76438-864 76438-866 76438-868 76438-870 76438-872 76438-874 76438-876 76438-878 76438-880 76438-882
High Performance FR Camo Jackets, Lakeland Industries
Workwear
Layer up and stay warm when donning the High-Performance FR Camo Jackets from Lakeland.
  • Inherently FR camo pattern fabric
  • Wind and water resistant
  • Four pocket design
  • Abrasion resistant finish
Outer layer garments complete the Layering System, giving wearers the comfort and protection required. Moisture wicking fabrics continue the process of pulling moisture away from the body to the outermost layer so that it can dry, while the wind-resistant and water-resistant fabrics help keep the elements at bay so wearers can focus on the task at hand.
Certifications: NFPA 70E. NFPA 2112 - UL certified. APTV: 14.6 cal/cm². CAT: 2.