igScript™ Probe-Based qPCR Master Mix, Intact Genomics
Supplier: INTACT GENOMICS, INC MS
4233 4235 4237
103853-770EA 356.92 CAD
103853-770 103853-772 103853-774
igScript™ Probe-Based qPCR Master Mix, Intact Genomics
Nucleic Acid Reagents RT-PCR and cDNA Reagents
igScript™ Probe-Based qPCR master mix provides improved PCR efficiency, wider dynamic range, superior sensitivity and specificity.
  • Enhanced efficiency, specificity, and sensitivity
  • Compatible with all real-time PCR instruments
  • Superior gene expression results under various cycling conditions
IgScript™ probe-based qPCR 2× master mix contains IgScript™ Taq DNA polymerase, MgCl₂, dNTPs, stabilizers, enhancers and low ROX reference dye with standard buffer providing improved qPCR efficiency, wider dynamic range, superior sensitivity and specificity. IgScript™ qPCR 2× master mix is a ready-to-use cocktail containing all components except primers, probe and template, for the amplification and detection of DNA in qPCR. This 2× master mix requires minimal handling during reaction setup and offer consistent and robust qPCR reactions. Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5´→3´ polymerase (1, 2) and a 5´→3´ exonuclease activity (3, 4). The amplification step features a high quality Taq DNA Polymerase which offers robust, reliable and better amplification.
Delivery information: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 pm will be shipped on the next business day. Orders received on Friday will be ship on the following Monday.
The shipping and dry ice packaging cost is $40 for your entire order in the 48 contiguous United States.