T5 Exonuclease
Supplier: New England Biolabs (NEB)
M0663L M0663S
CA76447-142EA 458 CAD
CA76447-142 CA76447-144
T5 Exonuclease
Enzymes
T5 Exonuclease degrades DNA in the 5' to 3' direction. T5 Exonuclease is able to initiate nucleotide removal from the 5' termini or at gaps and nicks of linear or circular dsDNA. However, the enzyme does not degrade supercoiled dsDNA. T5 Exonuclease also has ssDNA endonuclease activity.
  • Double-stranded DNA specific exonuclease and single-stranded DNA endonuclease
  • Initiates at the 5' termini of linear or nicked double-stranded DNA
  • Cleaves linear or nicked double-stranded DNA in the 5' to 3' direction
Order Now
SPECIFICATIONS
Enzyme name T5 Exonuclease
Environmentally Preferable
Source
Species
Concentration 10000 units/ml