HALO® Penta-HILIC, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
HALO®
95814-205 92812-505 95813-705 92813-605 92814-905 95812-105 98517-605 98215-605 98515-605 95811-905 98215-705 92813-405 92810-505 98217-705 98519-405 92813-205 95812-705 92811-705 95811-605 92811-405 95813-605 92812-905 92810-705 95811-305 98216-405 92811-605 95810-405 92811-305 98519-605 92813-505 91813-505 95814-505 95813-905 91812-705 98518-405 91813-205 91812-505 98517-705 98216-705 92812-605 92814-505 91813-405 98218-605 95810-505 98515-405 95812-205 95810-705 91813-605 91813-105 98218-405 92812-105 92814-305 95814-905 91813-305 98217-605 95813-205 98216-605 95814-405 98516-605 92813-105 92812-405 98517-405 98519-705 98516-705 95811-505 98518-605 98219-605 92812-705 91812-605 92813-705 98218-705 91813-705 91812-405 95813-305 95812-605 98515-705 92814-605 95810-905 92813-905 92814-705 95813-505 95811-405 95813-105 98516-405 95814-705 91812-105 95814-105 95814-605 92812-305 98215-405 92813-305 91812-305 91812-205 98219-705 95812-505 95813-405 98217-405 91813-905 95812-305 98219-405 92811-505 92814-405 92814-205 91812-905 92812-205 92810-405 95810-605 95814-305 95811-705 95812-905 95812-405 98518-705 92810-605 92814-105
76444-988EA 1441.31 CAD
76444-988 76445-790 76443-822 76445-430 76443-942 76445-550 76445-670 76443-422 76445-710 76445-830 76445-950 76443-418 76446-360 76446-480 76446-240 76446-520 76446-640 76446-400 76443-556 76443-432 76445-026 76444-580 76444-062 76445-810 76446-010 76444-620 76444-860 76444-928 76445-570 76444-800 76445-690 76445-970 76443-446 76443-842 76443-962 76445-450 76445-610 76443-322 76445-730 76445-850 76446-180 76446-580 76446-340 76446-460 76446-220 76446-500 76446-620 76443-450 76445-046 76443-576 76443-454 76444-082 76444-002 76444-948 76445-470 76445-590 76445-750 76445-870 76445-990 76443-862 76443-982 76445-510 76443-464 76445-630 76443-342 76446-160 76446-280 76446-560 76446-680 76446-320 76446-440 76444-672 76446-200 76446-720 76446-600 76443-478 76443-510 76443-596 76443-902 76444-820 76444-022 76443-482 76444-968 76445-490 76445-530 76445-650 76445-770 76443-402 76445-890 76445-930 76443-882 76443-486 76445-410 76444-692 76446-380 76446-140 76446-260 76446-540 76444-732 76446-660 76446-300 76446-420 76446-700 76444-712 76443-414 76443-410 76443-496 76445-910 76444-042 76443-406 76446-030 76443-922 76444-840 76444-600
HALO® Penta-HILIC, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® Penta-HILIC columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å Penta-HILIC columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
  • Ideal for polar compounds poorly retained in reversed-phase modes
  • Alternate selectivity for HILIC mode
  • Excellent peak shape for basic compounds in HILIC mode
  • Particle sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.