HALO® C4, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
HALO®
97219-614 92713-514 92712-314 92714-314 92710-714 92714-214 9271X-714 92711-614 92712-514 92712-214 92713-414 9271X-414 92714-414 92712-414 97219-714 92713-314 92712-614 97215-714 97218-414 92714-914 92710-514 92713-614 92710-614 92712-914 92713-114 92713-914 97218-614 9271X-614 97217-614 97215-614 92713-714 97216-714 97215-414 92710-414 92714-714 92713-214 92712-114 92714-514 97217-414 92712-714 97217-714 97219-414 97218-714 92711-514 97216-614 92714-114 92711-414 92711-714 92714-614 97216-414
76446-118EA 1524.59 CAD
76446-118 76444-506 76444-428 76443-386 76444-470 76444-392 76446-082 76444-434 76444-358 76446-088 76446-124 76446-046 76444-476 76444-398 76444-354 76443-392 76444-416 76537-458 76443-398 76446-094 76446-052 76444-380 76444-500 76446-130 76444-422 76444-348 76444-386 76446-058 76537-462 76444-342 76444-464 76537-466 76444-404 76444-494 76446-064 76446-100 76444-458 76444-452 76444-374 76444-410 76446-106 76446-070 76444-362 76444-482 76446-076 76444-368 76444-488 76444-446 76444-440 76446-112
HALO® C4, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS C4 1000 Å Protein columns made with innovative large pore Fused-Core® particle technology provide best-in-class performance for high-resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO® 1000 Å columns are optimized for high performance in HPLC, UHPLC and LCMS large molecule applications, delivering excellent reproducibility and column lifetimes.
  • Ideal for mAbs, ADCs and large proteins due to unrestricted access to the bonded phase
  • Best-in-class resolution for full characterization of mAbs and their variants
  • Exceptional mass transfer kinetics
  • Outstanding low bleed performance and high temperature stability
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.