HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
91122-102 91123-102 92122-102 92123-102 95122-102 95123-102 95124-102 91122-202 91122-302 91122-402 91122-502 91122-602 91122-702 91122-902 91123-302 91123-402 91123-502 91123-602 91123-702 91123-902 91225-402 91225-602 91225-702 91226-402 91226-602 91226-702 92120-402 92120-602 92120-702 92121-402 92121-602 92121-702 92121-902 92122-202 92122-302 92122-402 92122-502 92122-602 92122-702 92122-902 92123-202 92123-302 92123-402 92123-502 92123-602 92123-702 92123-902 92124-202 92124-302 92124-402 92124-502 92124-602 92124-702 92124-902 95120-402 95120-602 95120-702 95120-902 95122-202 95122-302 95122-402 95122-502 95122-602 95122-702 95122-902 95123-202 95123-302 95123-402 95123-502 95123-602 95123-702 95123-902 95124-202 95124-302 95124-402 95124-502 95124-602 95124-702 95124-902 9212X-402 9212X-602 9212X-702
76443-312PK 1124.96 CAD
76443-312 76443-314 76443-368 76443-372 76443-376 76443-538 76443-542 76443-546 76443-610 76443-612 76443-614 76443-616 76443-618 76443-620 76443-622 76443-624 76443-626 76443-628 76443-630 76443-632 76443-634 76443-696 76443-700 76443-704 76443-708 76443-712 76443-716 76443-720 76443-724 76443-728 76443-732 76443-736 76443-740 76443-744 76443-748 76443-752 76443-756 76443-760 76443-764 76443-768 76443-772 76443-776 76443-780 76443-784 76443-788 76443-792 76443-796 76443-800 76443-804 76443-808 76443-812 76444-216 76444-220 76444-224 76444-228 76444-232 76444-236 76444-240 76444-244 76444-248 76444-252 76444-256 76444-260 76444-264 76444-268 76444-272 76444-276 76444-280 76444-284 76444-288 76444-292 76444-296 76444-300 76444-304 76444-308 76444-312 76444-316 76444-320 76444-324 76444-328 76444-332 76444-336 76445-286 76445-290 76445-294 76445-298 76445-302 76445-304 76445-308 76445-312 76445-316 76445-320 76445-324 76445-326 76445-330 76445-334 76445-338 76445-342 76445-346 76445-350 76445-352 76445-356 76445-360 76445-364 76445-368 76445-372 76445-376 76445-378 76445-382 76445-386 76445-390 76445-394 76445-398 76445-402 76446-736 76537-452 76537-454 76537-456
HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS ES-C18 Peptide columns made with innovative Fused-Core® particle technology are ideal for both ultrafast- and ultrahigh-resolution separations of peptides and polypeptides up to 20 kDa. HALO® peptide columns offer comparable peak capacity to sub-2 μm non-core columns at 40 to 50% back pressure (2.7 μm), and 20% higher peak capacity than sub-2 μm non-core columns at comparable back pressure (2 μm). HALO 160 Å peptide columns are optimized for high performance in HPLC, UHPLC and LCMS peptide applications.
  • Ideal for tryptic digests, post-translational modifications (PTMs),
More Product Information