HALO® C4, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
93412-114 93413-114 93414-114 93410-414 93410-614 93410-714 93411-414 93411-714 93412-214 93412-314 93412-414 93412-514 93412-614 93412-714 93412-914 93413-214 93413-314 93413-414 93413-514 93413-614 93413-714 93413-914 93414-214 93414-314 93414-414 93414-514 93414-614 93414-714 93414-914 94315-414 94315-614 94315-714 94316-414 94316-614 94316-714
76443-524PK 1124.96 CAD
76443-524 76443-530 76443-534 76445-094 76445-098 76445-102 76445-106 76445-110 76445-114 76445-118 76445-122 76445-126 76445-130 76445-134 76445-138 76445-144 76445-148 76445-154 76445-160 76445-164 76445-168 76445-172 76445-176 76445-180 76445-184 76445-188 76445-192 76445-196 76445-200 76445-206 76445-210 76445-216 76445-222 76445-226 76445-230 76445-234 76445-238 76445-242 76445-246 76445-250 76445-254 76445-258 76445-262 76445-266 76445-270 76445-274 76445-278 76445-282 76446-742
HALO® C4, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS C4 400 Å Protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. These columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
  • Ideal for large protein analysis such as mAbs and polypeptides
  • Well suited for high speed protein separations
  • Outstanding low bleed performance and
More Product Information