HALO® Phenyl-Hexyl, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
92112-106 92113-106 92114-106 91215-406 91215-606 91215-706 91216-406 91216-606 91216-706 92110-406 92110-606 92110-706 92111-406 92111-606 92111-706 92112-206 92112-306 92112-406 92112-506 92112-606 92112-706 92112-906 92113-206 92113-306 92113-406 92113-506 92113-606 92113-706 92113-906 92114-206 92114-306 92114-406 92114-506 92114-606 92114-706 92114-906
76443-356PK 1124.96 CAD
76443-356 76443-360 76443-364 76443-638 76443-642 76443-646 76443-650 76443-654 76443-658 76443-662 76443-666 76443-670 76443-674 76443-678 76443-682 76443-686 76443-690 76443-694 76444-098 76444-102 76444-106 76444-110 76444-114 76444-118 76444-122 76444-126 76444-130 76444-132 76444-136 76444-140 76444-144 76444-148 76444-152 76444-156 76444-160 76444-164 76444-168 76444-172 76444-176 76444-180 76444-184 76444-188 76444-192 76444-196 76444-200 76444-204 76444-208 76444-212
HALO® Phenyl-Hexyl, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS Phenyl-Hexyl peptide columns made with innovative Fused-Core® particle technology are ideal for both ultrafast- and ultrahigh-resolution separations of peptides and polypeptides up to 20 kDa.
  • Ideal for tryptic digests, post-translational modifications (PTMs), polypeptides
  • HALO® Peptide columns can be used in series to increase peak capacity for UHPLC and HPLC analyses of complex tryptic digest samples
  • Particle Size: 2.7 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® peptide columns offer comparable peakMore Product Information