HALO® C8, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
HALO®
95812-908 95812-408 92812-708 91813-408 95811-608 98516-708 95813-608 95810-508 95814-708 95812-308 91813-908 95813-408 92812-408 91812-908 98215-608 98217-608 95812-208 92813-108 92814-608 95812-708 92814-108 95810-608 91812-508 92813-308 95813-208 98216-708 98218-708 91812-708 98215-408 98517-708 92812-308 92810-708 95812-108 91813-308 98216-408 91813-508 91812-308 91813-708 98515-408 92811-508 95810-708 92810-608 92812-608 92811-408 95812-508 92812-208 98517-608 98218-408 98215-708 92813-708 91813-108 92813-208 98217-408 98516-408 95813-308 98518-708 95814-908 92813-408 98515-608 98218-608 95812-608 92812-108 95814-508 95814-208 98217-708 98219-408 98516-608 95811-708 95813-508 98518-608 95814-608 92811-608 98519-608 91812-108 92813-608 95810-408 95814-308 92811-308 91812-208 92814-708 98219-708 92810-508 92813-508 91812-608 95810-908 95814-408 98519-708 92814-408 91812-408 92810-408 91813-608 95811-308 92812-908 95813-708 98519-408 98517-408 92814-308 95811-508 95813-108 92814-208 98216-608 95814-108 98518-408 92814-908 92811-708 98515-708 95811-408 95813-908 92813-908 95811-908 92812-508 91813-208 98219-608 92814-508
76444-866EA 1441.31 CAD
76444-866 76444-626 76445-032 76445-556 76445-676 76445-796 76444-994 76444-754 76446-366 76446-486 76446-246 76444-738 76446-526 76446-646 76446-406 76445-416 76443-828 76445-536 76445-656 76443-948 76445-816 76445-936 76446-016 76444-586 76444-068 76444-806 76444-008 76445-052 76443-602 76445-456 76445-576 76443-562 76445-696 76443-438 76446-186 76444-898 76444-534 76446-586 76444-770 76446-346 76446-466 76444-518 76446-226 76444-914 76446-506 76446-626 76445-956 76445-436 76443-848 76443-968 76445-716 76445-836 76443-328 76444-642 76444-088 76444-882 76444-826 76443-582 76445-596 76443-502 76443-348 76445-996 76445-476 76446-166 76444-678 76446-286 76444-550 76446-566 76446-686 76446-326 76446-446 76443-470 76446-206 76446-726 76444-658 76446-606 76444-934 76445-736 76445-856 76445-976 76443-868 76443-988 76445-616 76444-786 76444-028 76443-908 76444-606 76444-846 76443-888 76445-012 76445-496 76445-776 76445-896 76444-698 76446-386 76444-974 76446-146 76446-266 76446-546 76446-666 76446-306 76444-718 76446-426 76444-954 76446-706 76445-516 76445-636 76443-928 76445-756 76445-876 76445-916 76444-566 76444-048 76446-036
HALO® C8, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® C8 columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å C8 columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
  • Ideal for broad range of analytes
  • Less hydrophobic (less retentive) than C18
  • Good for ion-pair applications
  • Particle sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.