HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
92712-102 92713-102 92714-102 92710-402 92710-602 92710-702 92711-402 92711-602 92711-702 92712-202 92712-302 92712-402 92712-502 92712-602 92712-702 92712-902 92713-202 92713-302 92713-402 92713-502 92713-602 92713-702 92713-902 92714-202 92714-302 92714-402 92714-502 92714-602 92714-702 92714-902 97215-402 97215-602 97215-702 97216-402 97216-602 97216-702
76443-384PK 1124.96 CAD
76443-384 76443-390 76443-396 76444-340 76444-346 76444-352 76444-356 76444-360 76444-366 76444-372 76444-378 76444-384 76444-390 76444-396 76444-402 76444-408 76444-414 76444-420 76444-426 76444-432 76444-438 76444-444 76444-450 76444-456 76444-462 76444-468 76444-474 76444-480 76444-486 76444-492 76444-498 76444-504 76446-044 76446-050 76446-056 76446-062 76446-068 76446-074 76446-080 76446-086 76446-092 76446-098 76446-104 76446-110 76446-116 76446-122 76446-128
HALO® ES-C18, BIOCLASS, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® BIOCLASS ES-C18 1000 Å protein columns made with innovative large pore Fused-Core® particle technology provide best-in-class performance for high-resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns.
  • Ideal for mAbs, ADCs and large proteins due to unrestricted access to the bonded phase
  • Best-in-class resolution for full characterization of mAbs and their variants
  • Exceptional mass transfer kinetics
  • Outstanding low bleed performance and high temperature stability
HALO® 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.