HALO® RP-Amide, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
HALO®
95814-307 98515-407 95811-907 98216-707 91813-207 95811-407 92813-307 91812-407 98517-407 95810-607 98519-707 95812-107 92814-107 92814-307 95814-207 95811-707 91813-307 92814-207 92813-907 91812-307 98215-607 91813-407 92814-907 95811-607 98218-607 98217-707 91812-207 98218-707 92812-507 95813-607 92810-407 98219-607 92812-207 95813-207 98219-707 92811-507 95810-707 95812-707 98519-607 98216-407 92812-907 98515-707 95813-307 98516-607 95811-307 95812-407 95814-407 92814-507 92813-107 98516-407 98218-407 91813-507 98217-407 92812-607 98215-707 92812-707 91812-707 95814-507 95812-507 98517-607 95813-107 95813-907 91813-607 91813-907 92811-607 98216-607 92814-407 91812-107 92812-407 91813-707 98519-407 95813-507 95812-207 91812-607 92812-107 95810-907 92810-607 92813-707 92811-707 91812-507 92813-407 95812-307 92812-307 92813-607 92814-607 98518-407 95814-107 92813-507 98219-407 98215-407 92813-207 95810-507 95813-407 98217-607 95814-907 95810-407 95812-607 98516-707 95814-707 98518-607 95813-707 92810-707 95814-607 98515-607 92811-407 92814-707 95812-907 95811-507 92810-507 98518-707 92811-307 98517-707 91813-107 91812-907
76444-624EA 1347.31 CAD
76444-624 76445-030 76445-794 76445-434 76445-554 76445-674 76444-752 76446-364 76446-484 76446-244 76444-992 76446-524 76446-644 76446-404 76444-736 76443-436 76445-934 76445-414 76443-826 76443-946 76445-814 76444-864 76446-014 76444-584 76444-066 76443-560 76444-804 76444-768 76445-050 76445-574 76445-694 76443-326 76443-600 76445-974 76445-454 76446-184 76444-896 76444-532 76446-584 76444-656 76446-344 76446-464 76446-224 76446-504 76446-624 76444-912 76444-516 76445-714 76445-834 76445-954 76444-086 76443-846 76443-966 76444-006 76444-880 76444-640 76443-580 76444-548 76444-824 76443-866 76443-986 76445-474 76445-594 76443-468 76443-500 76443-346 76445-754 76445-874 76445-994 76446-164 76444-676 76446-284 76444-952 76446-564 76446-684 76446-324 76446-444 76446-204 76446-724 76444-932 76446-604 76445-614 76443-906 76445-734 76445-854 76444-026 76444-784 76444-844 76444-604 76443-886 76445-494 76445-010 76445-534 76445-654 76445-774 76445-894 76446-384 76444-972 76446-144 76446-264 76444-696 76446-544 76446-664 76446-304 76446-424 76444-716 76446-704 76445-514 76443-926 76445-634 76445-914 76446-034 76444-564 76444-046
HALO® RP-Amide, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® RP-Amide columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å RP-Amide columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
  • Ideal for basic compounds (alcohols, acids, phenols, catechins)
  • Alternate selectivity to alkyl phases
  • 100% aqueous compatible
  • Particle sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.