HALO® PFP, HPLC Columns, Advanced Materials Technology
Supplier: Advanced Materials Technology
HALO®
95810-409 91812-909 98217-609 91812-609 95814-709 95814-909 98516-609 92813-409 95810-609 95810-509 92814-409 91813-309 95814-409 95813-609 98515-409 91812-309 98217-709 92813-309 98517-709 92812-109 98216-409 95811-409 92810-409 95812-109 95811-609 92813-709 98519-609 91813-509 98215-709 95810-709 92813-109 91812-109 95813-909 95812-509 95812-709 95814-609 95811-909 92814-909 98518-409 98215-409 92811-409 98217-409 92814-709 91813-609 95814-309 95813-509 92810-709 92812-309 92811-609 95811-509 98516-409 98519-409 95812-309 95813-409 92813-909 91813-209 91813-409 91813-709 95812-409 92814-609 98519-709 95812-909 92812-909 92812-209 92810-609 98218-609 95810-909 98516-709 95812-209 98517-409 92811-709 95813-209 98215-609 92814-109 98218-409 98219-409 98216-609 91812-209 92812-709 91812-509 98219-609 95811-709 92813-609 92811-309 98517-609 95814-509 92812-409 95811-309 92814-209 98219-709 92813-509 91812-709 92811-509 91812-409 98515-609 95814-209 98518-609 91813-909 92814-309 98218-709 95813-709 92813-209 95813-109 92812-509 92812-609 95813-309 91813-109 98518-709 98515-709 98216-709 95814-109 92810-509 95812-609 92814-509
76444-628EA 1347.31 CAD
76444-628 76444-900 76444-868 76445-034 76445-798 76444-070 76444-996 76446-368 76446-488 76446-248 76446-528 76444-976 76446-648 76446-408 76443-830 76443-950 76445-538 76445-658 76445-778 76445-898 76445-938 76445-418 76444-588 76445-818 76444-740 76443-440 76444-090 76445-054 76445-578 76443-564 76445-698 76444-808 76444-010 76446-188 76446-588 76444-772 76443-330 76446-348 76446-468 76444-916 76446-228 76444-756 76446-508 76446-628 76443-850 76443-970 76445-438 76445-558 76445-678 76445-718 76445-838 76445-958 76443-604 76444-884 76444-644 76444-520 76444-828 76445-478 76445-598 76443-584 76444-030 76446-168 76446-288 76444-552 76443-990 76446-568 76443-472 76446-688 76444-936 76446-328 76443-350 76446-448 76446-208 76444-536 76446-728 76443-910 76446-608 76443-870 76445-978 76445-458 76445-618 76445-738 76445-858 76443-504 76444-700 76444-788 76444-660 76446-018 76444-848 76444-608 76445-014 76445-498 76444-050 76446-388 76446-148 76446-268 76443-890 76446-548 76446-668 76446-308 76446-428 76444-956 76446-708 76443-930 76445-758 76445-878 76445-998 76444-680 76445-518 76445-638 76444-720 76444-568 76445-918 76446-038
HALO® PFP, HPLC Columns, Advanced Materials Technology
Chromatography Columns
HALO® PFP columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å PFP columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
  • Ideal for aromatics and electron-rich compounds
  • Alternate selectivity to alkyl phases
  • Useful in reversed-phase and HILIC modes
  • Particle sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.