Syringe Filters, Non Sterile, Environmental Express®
Supplier: Environmental Express
SF012G SF015G SF020CA SF020E SF020N SF020T SF020V SF045CA SF045E SF045N SF045T SF045V SF1070G SF120E SF120N SF120T SF120V SF145CA SF145E SF145N SF145T SF145V
76437-128PK 474.27 CAD
76437-128 76437-130 76437-132 76437-134 76437-136 76437-138 76437-140 76437-142 76437-144 76437-146 76437-148 76437-150 76437-152 76437-154 76437-156 76437-158 76437-160 76437-162 76437-164 76437-166 76437-168 76437-170
Syringe Filters, Non Sterile, Environmental Express®
Filters Syringe Filters
Economical, single use, and efficient filtering stream for high output filtration from viscous and particle-laden solutions
  • Reduce bursting with reinforced polypropylene housing
  • Take advantage of a large effective filtration area
  • Standard female Luer-lock inlet and male Luer-slip outlet
  • Made with USP Class VI components to ensure quality
  • Available in various membrane size, pore size, and material combinations
Use single-use syringe filters for clarification and prefiltration, and even for removing fine particulate with the membrane's large effectiveMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable