HotBlock® Watch Glasses and Reflux Caps, Environmental Express®
Supplier: Environmental Express
SC417 SC505 SC506
76437-112PK 295.11 CAD
76437-112 76437-118 76437-120
HotBlock® Watch Glasses and Reflux Caps, Environmental Express®
Watch Glasses
Choose from reflux caps or watch glasses compatible with digestion cups.
  • Optimize use of your HotBlock® digestion cup
These disposable 34 mm reflux caps fit onto our 50 ml digestion cups. This special cap has a 12.7 mm hole in its center to allow for the addition of reagents during digestion.
The watch glasses feature a ribbed design which allows partial reflux during digestion. Watch glass has a large center stem to aid in handling.