FlipMate™ Filtration Assemblies, Environmental Express®
Supplier: Environmental Express
SC0301 SC0302 SC0303 SC0304 SC0306 SC0308 SC0309 SC0602 SC0603 SC0604 SC0605 SC0606 SC0607 SC0608 SC0651 SC0652
76436-996PK 248.27 CAD
76436-996 76437-000 76437-004 76437-008 76437-012 76437-016 76437-020 76437-038 76437-042 76437-046 76437-050 76437-054 76437-056 76437-058 76437-060 76437-062
FlipMate™ Filtration Assemblies, Environmental Express®
Filters Membrane Filter Units
The FlipMate™ assemblies easily filter your sample with the use of two threaded digestion cups, a little tubing, and vacuum pressure.
  • Sample is ready in just seconds
  • Convenient, economical method for filtration
  • Produce quality results with clean samples
The sample is placed in one cup, the filter assembly threaded onto the top of the cup, then the cup and filter are flipped, the receiving cup attached, vaccum tubing attached, vaccum pressure applied, and the sample is then pulled from the sample cup, through the filter assembly, and into the empty cup.
Ordering information: Digestions cups, tubing, and vacuum source are not included, filtration assembly only.