Washed and Dried TSS/VSS Glass Fiber Filters, Environmental Express®
Supplier: Environmental Express
F924110MM F924110VOL F924125MM F92424MM F92432MM F92432VOL F92434MM F92447VOL F92455MM F92470MM F92490MM F92490VOL
76436-888PK 242.25 CAD
76436-888 76436-890 76436-892 76436-894 76436-896 76436-898 76436-900 76436-902 76436-904 76436-906 76436-908 76436-910
Washed and Dried TSS/VSS Glass Fiber Filters, Environmental Express®
Filters Glass Fiber Filters
Ready to be weighed and used for suspended solids filtration.
  • Use in Standard Methods 2540
  • Provide consistently stable weights
  • Made from binderless borosilicate glass and have a 1.5 µm pore size
Laser cutting ensures a clean, sealed, fiber-free edge that maintains its integrity throughout washing and drying, as well as your testing process. These filters are rinsed three times with DI water to ensure they are fiber free. TSS filters are then dried in an oven at 105 °C to obtain stable weights. VSS filters are dried at 550 °C to ensure all volatiles are removed.