ProWeigh® and ProWeigh® Double Filters, Environmental Express®
Supplier: Environmental Express
F934110MM F934110VOL F93425MM F93425VOL F93432MM F93442MM F93442VOL F93447MM F93447MM-X F93447VOL F93447VOL-X F93455MM F93455VOL F93470MM F93470VOL F93476MM-X F93476VOL F93476VOL-X F93490MM-X F93490VOL F93490VOL-X
76436-912PK 423.98 CAD
76436-912 76436-914 76436-916 76436-918 76436-920 76436-922 76436-924 76436-926 76436-928 76436-930 76436-932 76436-934 76436-936 76436-938 76436-940 76436-942 76436-944 76436-946 76436-948 76436-950 76436-952
ProWeigh® and ProWeigh® Double Filters, Environmental Express®
Filters Glass Fiber Filters
These ready-to-use filters for total and volatile suspended solids decrease sample prep time for your suspended solids testing.
  • Approved for Standard Methods 2540
  • Each filter is individually prepared, weighed, and labeled to be ready for use upon receipt
  • Eliminate loose fibers and inaccurate weights with precise, laser-cut edges
  • Quality control ensures filter integrity with visual inspection after each cutting, washing, and drying step
  • Made from binderless borosilicate glass
All ProWeigh filters are prepared according to Standard Methods requirements. Filters are weighed, washed 3 times in deionized water, dried at 105 °C and weighed to 0.1 mg. Filters come packaged in a labeled aluminum pan with the weight of the filter printed on the label. In addition, the filter weight and ID number are printed in code 128 bar coding.
Delivery information: Supplied with TKN100 base controller, 28-or 54-well TKN HotBlock, choice of 28 or 54× threaded (C6010) or non-threaded tubes (TKN6011), plus 28 or 54× teardrop stoppers (TKN6012), depending on options selected, see description below for full details.