Potassium thiocyanate buffered solution, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = R

BAKR3329-09 BAKR3329-TG
CAJT3329-09CS 63125.74 CAD
CAJT3329-09 CAJT3329-TG
Potassium thiocyanate buffered solution, BAKER ANALYZED®, J.T.Baker®
Potassium thiocyanate
Formula: KSCN
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (clear, colorless solution) Passes Test
Potassium Thiocyanate (KSCN), g/100 mL 55.3 - 63.6
Sodium (Na), g/L 2.6 - 3.6
Potassium (K), mEQ/L 5414 - 6616
pH (As Is" basis) at 25*C" 6.8 - 7.3
Chloride Identity (Cl) Passes Test
Phosphate Identity (PO₄) Passes Test
Sterility Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR