Monarch® RNA Cleanup Columns (50 µg), New England Biolabs
Supplier: New England Biolabs
Monarch®
T2047L T2047L
CA76299-642EA 378 CAD
CA76299-642 76299-642
Monarch® RNA Cleanup Columns (50 µg), New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Monarch® RNA Cleanup Columns (50 µg) are a component of the Monarch® RNA Cleanup Kit (50 µg) and can be used to purify up to 50 µg of RNA from enzymatic reactions. 
  • Ideal for the cleanup of RNA from enzymatic reactions including sgRNA synthesis, DNase I treatment, labeling, and capping.
  • Elute in ≥20 µl for concentrated RNA
  • 70 to 100% RNA recovery
  • Unique column design prevents buffer carryover and elution of silica particulates
  • Purified RNA is ready for use in a wide variety of downstream applications
The columns are ideal for cleaning up RNA after in vitro transcription reactions such as sgRNA synthesis reactions. The columns are designed without the use of a retaining ring, ensuring no buffer retention and no carryover of contaminants. RNA can be eluted in as little as 20 μl for and is ready for use in a variety of downstream applications including RT-PCR, RNA Library Prep for NGS and transfection.
Monarch® RNA Cleanup Columns are also available for 10 and 500 µg binding capacities.