Socorex® Qualitix® Sterilization Pouches, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
3210.0816 3210.0926 3210.1326 3210.1936 3210.3040
76437-376CS 32.76 CAD
76437-376 76437-378 76437-380 76437-382 76437-384
Socorex® Qualitix® Sterilization Pouches, DWK Life Sciences
Bags Sterilization Pouches
The Qualitix® pouches provide easy and effective wrapping of items requiring sterilization.
  • Robust durable material
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
Fitting any small to mid-size laboratory device, the Qualitix® single use pouches provide easy and effective wrapping of items requiring sterilization. Made with robust medical grade paper, the pouches tightly close with a self-sealing strip to maintain sterility. ContentMore Product Information