HotBlock® 200 Digestion Heating Blocks, Environmental Express®
Supplier: Environmental Express
SC2015-96 SC2015-96V240 SC2050-36 SC2050-36V240 SC2050-54 SC2050-54V240 SC2100-25 SC2100-25V240 SC2100-35 SC2100-35V240
76437-088EA 9209.17 CAD
76437-088 76437-090 76437-092 76437-094 76437-096 76437-098 76437-100 76437-102 76437-104 76437-106
HotBlock® 200 Digestion Heating Blocks, Environmental Express®
Digestion Systems Mineralisation Workstations
Same dependability with increased temperature range.
  • Reach higher temperatures—perform more procedures
  • PTFE-coated blocks for chemical and corrosion resistance
  • Included polycarbonate transfer racks set on the block for easy placement and removal
Perform a wider range of experiments and digestions with the expanded temperature range—up to 200 °C* with the new HotBlock 200! Blocks are available with 25 to 96 wells, accepting cup sizes of 15, 50, or 100 ml, and controllers accept one or two block heaters.
Our blocks continue to be constructed of metals-free materials to avoid cross contamination and provide chemical resistance. The block housing is made of durable Kydex, and the block is constructed of PTFE-coated graphite. Polycarbonate racks allow for easy insertion and removal of multiple digestion cups.
Ordering information: Blocks and controllers are sold separately.