2-8CELLsius Short-Term Storage Solutions, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
PP338-100
PP338-500
PP338-1000
PP338-1000B
76443-304EA
200.63
CAD
76443-304
76443-306
76443-308
76443-310
2-8CELLsius Short-Term Storage Solutions, Protide Pharmaceuticals
Cell Culture Supplements and Additives
Ready to use, balanced, cytoprotective, protein free solution with bio-stabilizers for short-term storage, regenerative medicine, stem cell processing and transportation of biological material. Short term storage can be achieved at 2-8°C or at ambient temperature for 24-48 hours. Does not contain any growth factors, hormones, or antibiotics. 2-8CELLsius™ can also be utilized as a cell washing solution.
- TYPE II drug master file
- USP grade components/Highest grade
- Supports high cell viability
- No animal or human proteins
- No growth factors, hormones, antibiotics
- Ready to use/sterile membrane filtered
Certifications: cGMP manufactured.