2-8CELLsius + DMSO Defined Cryopreservation Solutions, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
PP338DM5-10 PP338DM5-50 PP338DM5-100 PP338DM10-10 PP338DM10-50 PP338DM10-100 338DM10-1000B 338DM20-1000B PP338DM5-100B PP338DM10-100B PP338DM20-100B
76443-282EA 43.01 CAD
76443-282 76443-284 76443-286 76443-288 76443-290 76443-292 76443-294 76443-296 76459-572 76459-574 76459-576
2-8CELLsius + DMSO Defined Cryopreservation Solutions, Protide Pharmaceuticals
Cell Culture Supplements and Additives
When combined with DMSO, 2-8CELLsius™ can be utilized as a cryopreservation solution. 2-8CELLsius™ + DMSO is a cytoprotective, protein free solution, with bio-stabilizers that support high post thaw cell viability. Optimal recovery is achieved through a novel approach to cell physiology at ultra-low temperatures.
  • TYPE II drug master file
  • USP grade components/highest grade
  • Supports high cell viability
  • No animal or human proteins
  • No growth factors, hormones, antibiotics
  • Ready to use/sterile membrane filtered
Certifications: cGMP manufactured.