TCH Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
3001
3002
3005
76443-264EA
273.4
CAD
76443-264
76443-266
76443-268
TCH Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Cell Culture Supplements and Additives
Serum replacement used primarily for human cells and the production of cell secreted proteins. TCH™ is also effective in culturing other mammalian cells without the use of animal proteins. Contains only human proteins (i.e., human serum albumin, human transferrin, recombinant insulin), which are heat treated.
- Defined, serum-free, xeno-free
- No hormones, antibiotics, growth factors, phenol red, Ig
- 50X concentrate
- Can be utilized with a wide variety of basal media
Certifications: cGMP manufactured.