TM-235 Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
2001 2002 2005
76443-258EA 225.17 CAD
76443-258 76443-260 76443-262
TM-235 Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Cell Culture Supplements and Additives
Serum replacement with similar properties to TCM™ but contains additional components for cell types that have required higher concentrations of serum (typically more than 10%). TM-235™ is defined and multi-purpose. Contains highly purified, heat-treated bovine serum albumin, heat-treated bovine transferrin and recombinant insulin.
  • Defined, serum-free
  • No hormones, antibiotics, growth factors, phenol red, Ig
  • 50X concentrate
  • Can be utilized with a wide variety of basal media
Certifications: cGMP manufactured.