TCM Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Supplier: PROTIDE PHARMACEUTICALS
1001
1002
1005
76443-252EA
192.98
CAD
76443-252
76443-254
76443-256
TCM Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Cell Culture Supplements and Additives
General serum replacement for many cell types from a variety of species. TCM™ is a defined, multi-purpose serum replacement useful in studying the effects of growth factors and the production of proteins. TCM™ contains highly purified, heat-treated bovine serum albumin, heat-treated bovine transferrin and recombinant insulin.
- Defined, serum-free
- No hormones, antibiotics, growth factors, phenol red, Ig
- 50X concentrate
- Can be utilized with a wide variety of basal media
Certifications: cGMP manufactured.