Revision 4.0 Add-Ons Mix for US EPA Method 524.2, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
ECS-A-046XP ECS-B-046XP
SXECS-A-046XPEA 176.58 CAD
SXECS-A-046XP SXECS-B-046XP
Revision 4.0 Add-Ons Mix for US EPA Method 524.2, SPEX CertiPrep
Standards Environmental Analysis Standards
ECS Standards for GC/MS
All ampulized standards are supplied with a pre-labeled transfer/storage vial. We want to make the transfer of ampulized standards as easy as possible. The label includes the catalog number, lot number, product description and expiration date.