9 Regulated Phthalates and BPA Standard in Medium Density Polyethylene, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
CRM-PE002
SXCRM-PE002EA 446.71 CAD
SXCRM-PE002
9 Regulated Phthalates and BPA Standard in Medium Density Polyethylene, SPEX CertiPrep
Standards Environmental Analysis Standards
Consumer safety standards, phthalates in polyethylene QC standards.
Polyethylene (PE) is one of the world’s most common plastics. PE is used in a variety of common consumer products including children’s toys and care items. Current US regulations limit the concentrations of certain phthalates in childcare articles and children’s toys. Laboratories are tasked with the analysis of children’s toys for these potentially hazardous phthalates.