Plastic Face Shield, Bal Supply
Supplier: Bal Supply
PFS
76443-194CS 460.29 CAD
76443-194
Plastic Face Shield, Bal Supply
Face Shields
Premium quality face shield features a clear PET lens designed to protect the face from accidental splashes.
  • PET lens, 14 mil thickness
  • Anti-fog and anti-static coating
  • Comfortable foam head piece
  • Single-use design
  • Latex-free
  • Nonsterile
Premium quality, optically clear, distortion-free lens. Face shields feature a foam head piece and soft, stretchable headband for comfortable use over extended periods.
Packaging: Face shields are individually bagged, and supplied 200 per case.